Product Details

SNP ID
rs12699799
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:16461981 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGGGTCAGCATGCAGGGTATAATTT[C/T]ATACTATGCGACCGTAAAGAGCTAC
Phenotype
MIM: 609675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOSTDC1 PubMed Links

Gene Details

Gene
SOSTDC1
Gene Name
sclerostin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015464.2 1369 UTR 3 NP_056279.1
XM_017011943.1 1369 UTR 3 XP_016867432.1

View Full Product Details