Product Details
- SNP ID
-
rs17230081
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:114330749 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGATGGGCGATTGGCCACTTCTCCT[C/T]GATAACATTACTGTTTTTCTTCCGC
- Phenotype
-
MIM: 605729
MIM: 138600
MIM: 138610
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKNA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116608376] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKNA
- Gene Name
- AT-hook transcription factor
There are no transcripts associated with this gene.
- Gene
- ORM1
- Gene Name
- orosomucoid 1
There are no transcripts associated with this gene.
- Gene
- ORM2
- Gene Name
- orosomucoid 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000608.2 |
|
Intron |
|
|
NP_000599.1 |
View Full Product Details