Product Details
- SNP ID
-
rs7031239
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:122552526 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGAAGGGAGAGAGTGGCTCCCTGT[C/T]CTATTTGTATGCGTGTGTGTGGGAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR1L8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75768770] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR1L8
- Gene Name
- olfactory receptor family 1 subfamily L member 8
- Gene
- OR1N2
- Gene Name
- olfactory receptor family 1 subfamily N member 2
View Full Product Details