Product Details

SNP ID
rs11769057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100367038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTAGGGCGCAGCAGAGAAGGTGGG[A/G]TGGAGACTGCTATCACCTGGATGCC
Phenotype
MIM: 605341 MIM: 605342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
There are no transcripts associated with this gene.

Gene
PILRB
Gene Name
paired immunoglobin-like type 2 receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178238.3 Intron NP_839956.1
Gene
STAG3L5P-PVRIG2P-PILRB
Gene Name
STAG3L5P-PVRIG2P-PILRB readthrough
There are no transcripts associated with this gene.

View Full Product Details