Product Details

SNP ID
rs17544690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77181676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTACATTCATAGGAACTCTTGATA[C/T]ATTTAGAAAGTAGGCCTTTGTGTAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC146 PubMed Links

Gene Details

Gene
CCDC146
Gene Name
coiled-coil domain containing 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020879.2 Intron NP_065930.2

View Full Product Details