Product Details

SNP ID
rs2005084
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:37884240 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCATTAGTCAATTGGAAGTGGCAC[A/G]AGTTTCATCTGGAAAAAATATTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A51 PubMed Links

Gene Details

Gene
SLC25A51
Gene Name
solute carrier family 25 member 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033412.3 Intron NP_219480.1

View Full Product Details