Product Details

SNP ID
rs7005125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:19316519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAAGCAATGTCCACACCTGGG[C/T]CAGTGTGACAGTCGGTTCCTTGCTG
Phenotype
MIM: 614968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH2D4A PubMed Links

Gene Details

Gene
SH2D4A
Gene Name
SH2 domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174159.1 Intron NP_001167630.1
NM_001174160.1 Intron NP_001167631.1
NM_022071.3 Intron NP_071354.2
XM_011544610.1 Intron XP_011542912.1
XM_017013729.1 Intron XP_016869218.1

View Full Product Details