Product Details

SNP ID
rs11993596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38514422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAATTCTCAGTTTTAAAATAAA[C/T]ACCTGGGGCTGCTTGTAACTTTTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf86 PubMed Links

Gene Details

Gene
C8orf86
Gene Name
chromosome 8 open reading frame 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303531.1 Intron NP_001290460.1
NM_207412.2 Intron NP_997295.1

View Full Product Details