Product Details

SNP ID
rs11587982
Assay Type
Validated
NCBI dbSNP Submissions
26
Location
Chr.1:45696196 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAATGCTATTTTTCCAGGTGAATT[A/T]CAGTGTATGCTTCTTTAAGAAAACT
Phenotype
MIM: 147485
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GPBP1L1 PubMed Links

Gene Details

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
IPP
Gene Name
intracisternal A particle-promoted polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145349.1 5571 Intron NP_001138821.1
NM_005897.2 5571 Intron NP_005888.1
XM_005270839.4 5571 UTR 3 XP_005270896.1
XM_006710623.3 5571 UTR 3 XP_006710686.1
XM_011541394.2 5571 Intron XP_011539696.1
XM_017001210.1 5571 Intron XP_016856699.1
Gene
TMEM69
Gene Name
transmembrane protein 69
There are no transcripts associated with this gene.

View Full Product Details