Product Details

SNP ID
rs12565895
Assay Type
Validated
NCBI dbSNP Submissions
40
Location
Chr.1:62597506 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTGGTATATATAGAGTTAAGAAGT[C/T]TAGGTCTGCTTCCAGAAGAAAACAG
Phenotype
MIM: 604774 MIM: 615730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGPTL3 PubMed Links

Gene Details

Gene
ANGPTL3
Gene Name
angiopoietin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014495.3 20 UTR 5 NP_055310.1
Gene
DOCK7
Gene Name
dedicator of cytokinesis 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271999.1 20 Intron NP_001258928.1
NM_001272000.1 20 Intron NP_001258929.1
NM_001272001.1 20 Intron NP_001258930.1
NM_001272002.1 20 Intron NP_001258931.1
NM_033407.3 20 Intron NP_212132.2
XM_005271292.2 20 Intron XP_005271349.1
XM_011542326.2 20 Intron XP_011540628.1
XM_011542327.2 20 Intron XP_011540629.1
XM_011542328.2 20 Intron XP_011540630.1
XM_011542330.2 20 Intron XP_011540632.1
XM_017002639.1 20 Intron XP_016858128.1
XM_017002640.1 20 Intron XP_016858129.1

View Full Product Details