Product Details

SNP ID
rs4429311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11826089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAAGATCACTATTTACACAATGG[C/G]AAGTAATTATGTCTTTGCAACTGTA
Phenotype
MIM: 613746 MIM: 615874
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCAR4 PubMed Links

Gene Details

Gene
BCAR4
Gene Name
breast cancer anti-estrogen resistance 4 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RSL1D1
Gene Name
ribosomal L1 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details