Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004965.6 | 639 | Intron | NP_004956.5 | ||
XM_011529543.1 | 639 | Intron | XP_011527845.1 | ||
XM_011529544.1 | 639 | Intron | XP_011527846.1 | ||
XM_011529545.1 | 639 | Intron | XP_011527847.1 | ||
XM_011529547.1 | 639 | Intron | XP_011527849.1 | ||
XM_011529549.1 | 639 | Intron | XP_011527851.1 | ||
XM_017028332.1 | 639 | Intron | XP_016883821.1 | ||
XM_017028333.1 | 639 | Intron | XP_016883822.1 | ||
XM_017028334.1 | 639 | Intron | XP_016883823.1 | ||
XM_017028335.1 | 639 | Intron | XP_016883824.1 | ||
XM_017028336.1 | 639 | Intron | XP_016883825.1 | ||
XM_017028338.1 | 639 | UTR 3 | XP_016883827.1 |