Product Details

SNP ID
rs9915183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:54969776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTATTTATTGCCCTCTCAACTGT[A/G]GGCCTGACACCAATCGACCTGCTGG
Phenotype
MIM: 603648 MIM: 610415 MIM: 604701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX11 PubMed Links

Gene Details

Gene
COX11
Gene Name
COX11, cytochrome c oxidase copper chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162861.2 1249 Intron NP_001156333.1
NM_001162862.2 1249 Intron NP_001156334.1
NM_001321518.1 1249 Intron NP_001308447.1
NM_004375.4 1249 Intron NP_004366.1
XM_011524342.2 1249 UTR 5 XP_011522644.1
XM_017024192.1 1249 UTR 5 XP_016879681.1
XM_017024193.1 1249 Intron XP_016879682.1
XM_017024194.1 1249 Intron XP_016879683.1
XM_017024195.1 1249 Intron XP_016879684.1
XM_017024196.1 1249 Intron XP_016879685.1
Gene
STXBP4
Gene Name
syntaxin binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178509.5 1249 Intron NP_848604.3
XM_005257187.4 1249 Intron XP_005257244.1
XM_006721797.3 1249 Intron XP_006721860.1
XM_006721798.3 1249 Intron XP_006721861.1
XM_017024410.1 1249 Intron XP_016879899.1
XM_017024411.1 1249 Intron XP_016879900.1
XM_017024412.1 1249 Intron XP_016879901.1
XM_017024413.1 1249 Intron XP_016879902.1
XM_017024414.1 1249 Intron XP_016879903.1
XM_017024415.1 1249 Intron XP_016879904.1
Gene
TOM1L1
Gene Name
target of myb1 like 1 membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details