Product Details

SNP ID
rs9803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18805296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAAAAACTATTAGCCTGTTTT[C/G]AAAGAAAAACATTCTAAAAGTGTGC
Phenotype
MIM: 607669 MIM: 607032
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL6IP1 PubMed Links

Gene Details

Gene
ARL6IP1
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
There are no transcripts associated with this gene.

Gene
SMG1
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015092.4 13579 UTR 3 NP_055907.3
XM_005255182.3 13579 UTR 3 XP_005255239.1
XM_005255183.3 13579 UTR 3 XP_005255240.1
XM_005255184.3 13579 UTR 3 XP_005255241.1
XM_011545769.2 13579 UTR 3 XP_011544071.1
XM_011545770.1 13579 UTR 3 XP_011544072.1
XM_017023066.1 13579 UTR 3 XP_016878555.1
XM_017023067.1 13579 UTR 3 XP_016878556.1

View Full Product Details