Product Details

SNP ID
rs7153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121664450 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGTGAGAGAATTCCAAGTTTTGA[A/G]GGGAGTGGTCCAAAGAGTAACAACT
Phenotype
MIM: 602500 MIM: 601306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLGB1 PubMed Links

Gene Details

Gene
GOLGB1
Gene Name
golgin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256486.1 10315 UTR 3 NP_001243415.1
NM_001256487.1 10315 UTR 3 NP_001243416.1
NM_001256488.1 10315 UTR 3 NP_001243417.1
NM_004487.4 10315 UTR 3 NP_004478.3
XM_005247371.4 10315 UTR 3 XP_005247428.1
XM_005247372.4 10315 UTR 3 XP_005247429.1
XM_005247373.1 10315 UTR 3 XP_005247430.1
XM_006713587.1 10315 UTR 3 XP_006713650.1
XM_006713588.1 10315 UTR 3 XP_006713651.1
XM_006713589.1 10315 UTR 3 XP_006713652.1
XM_006713590.1 10315 UTR 3 XP_006713653.1
XM_006713591.1 10315 UTR 3 XP_006713654.1
XM_011512699.2 10315 UTR 3 XP_011511001.1
XM_017006189.1 10315 UTR 3 XP_016861678.1
XM_017006190.1 10315 UTR 3 XP_016861679.1
XM_017006191.1 10315 UTR 3 XP_016861680.1
XM_017006192.1 10315 UTR 3 XP_016861681.1
XM_017006193.1 10315 UTR 3 XP_016861682.1
XM_017006194.1 10315 UTR 3 XP_016861683.1
XM_017006195.1 10315 UTR 3 XP_016861684.1
Gene
HCLS1
Gene Name
hematopoietic cell-specific Lyn substrate 1
There are no transcripts associated with this gene.

View Full Product Details