Product Details

SNP ID
rs1859012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:9789825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAAAAAGAATGCCAGAAGCCT[G/T]AGCCCGAAGCTCCAAAGAAGACAGT
Phenotype
MIM: 300103
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SHROOM2 PubMed Links

Gene Details

Gene
SHROOM2
Gene Name
shroom family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320663.1 Intron NP_001307592.1
NM_001320664.1 Intron NP_001307593.1
NM_001649.3 Intron NP_001640.1
XM_005274500.4 Intron XP_005274557.2
XM_011545506.1 Intron XP_011543808.1
XM_017029492.1 Intron XP_016884981.1
XM_017029493.1 Intron XP_016884982.1
XM_017029494.1 Intron XP_016884983.1

View Full Product Details