Product Details

SNP ID
rs648710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126306481 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAGGAAATCCCAAGTATTGGGA[A/T]TTCAAAGGCCCTGGGAGCTTCTGTC
Phenotype
MIM: 610534
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCPS PubMed Links

Gene Details

Gene
DCPS
Gene Name
decapping enzyme, scavenger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014026.4 Intron NP_054745.1
XM_011542778.2 Intron XP_011541080.1
XM_011542780.2 Intron XP_011541082.1
XM_017017612.1 Intron XP_016873101.1
Gene
HP11014
Gene Name
uncharacterized HP11014
There are no transcripts associated with this gene.

View Full Product Details