Product Details

SNP ID
rs1914407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:190975431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAATGACATTTCCAAAATTTTTT[C/T]TACCTTTTATAAAATGAAACAACAA
Phenotype
MIM: 138280 MIM: 600555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLS PubMed Links

Gene Details

Gene
GLS
Gene Name
glutaminase
There are no transcripts associated with this gene.

Gene
STAT1
Gene Name
signal transducer and activator of transcription 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007315.3 Intron NP_009330.1
NM_139266.2 Intron NP_644671.1

View Full Product Details