Product Details

SNP ID
rs941048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:18081322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTGATAATTCTGATAAACATCA[C/T]TTCCACCTCCAGAGACTGCTCTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RERGL PubMed Links

Gene Details

Gene
RERGL
Gene Name
RERG like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286201.1 694 Missense Mutation ATG,GTG M162V NP_001273130.1
NM_024730.3 694 Missense Mutation ATG,GTG M163V NP_079006.1

View Full Product Details