Product Details
- SNP ID
-
rs941047
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:18081163 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTGAATGTTTGAAACTCTCTGATA[C/T]AGGAAATCTCCCAGGATTACCTGTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RERGL
PubMed Links
Gene Details
- Gene
- RERGL
- Gene Name
- RERG like
View Full Product Details