Product Details

SNP ID
rs3814828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92940357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCCCCAACCCTTTTCTCTGC[A/G]GAGGGGGCTGCCTGGATCAGGGGTC
Phenotype
MIM: 118910 MIM: 601838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHGA PubMed Links

Gene Details

Gene
CHGA
Gene Name
chromogranin A
There are no transcripts associated with this gene.

Gene
ITPK1
Gene Name
inositol-tetrakisphosphate 1-kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017021262.1 2448 UTR 3 XP_016876751.1
XM_017021263.1 2448 UTR 3 XP_016876752.1
XM_017021264.1 2448 UTR 3 XP_016876753.1

View Full Product Details