Product Details

SNP ID
hCV11926455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113110749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCGGCGCCGGCGCGCCAACGCG[C/T]TCCTGGAGGAGCTGCGGCCGGGCTC
Phenotype
MIM: 613878
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F7 PubMed Links

Gene Details

Gene
F7
Gene Name
coagulation factor VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000131.4 178 Missense Mutation CTC,TTC L64F NP_000122.1
NM_001267554.1 178 Intron NP_001254483.1
NM_019616.3 178 Missense Mutation CTC,TTC L42F NP_062562.1

View Full Product Details