Product Details

SNP ID
rs137852336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154532625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACTCCTCGGGGTTGAAGAACATG[T/C]CCGGCTTCTTGGTCATCATCTTGGT
Phenotype
MIM: 305900 MIM: 300248
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
G6PD PubMed Links

Gene Details

Gene
G6PD
Gene Name
glucose-6-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000402.4 1356 Missense Mutation GAC,GGC D440G NP_000393.4
NM_001042351.2 1356 Missense Mutation GAC,GGC D410G NP_001035810.1
Gene
IKBKG
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
There are no transcripts associated with this gene.

View Full Product Details