Product Details
- SNP ID
-
rs1865951
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:235506695 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGGAGCCACCGGAAGCCGGGTCT[A/G]TATGGGCAGGGCCAGTGAGATGCAA
- Phenotype
-
MIM: 608651
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76603796] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGAP1
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
- Gene
- AGAP1-IT1
- Gene Name
- AGAP1 intronic transcript 1
There are no transcripts associated with this gene.
View Full Product Details