Product Details

SNP ID
rs9167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35009140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTATTGAAACTTTATTACAAA[A/G]TTATAAAGCAGAGCTCTGTAACAAA
Phenotype
MIM: 600940 MIM: 609735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIG3 PubMed Links

Gene Details

Gene
LIG3
Gene Name
DNA ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002311.4 7761 Intron NP_002302.2
NM_013975.3 7761 Intron NP_039269.2
XM_005257970.3 7761 UTR 3 XP_005258027.1
XM_006721896.3 7761 Intron XP_006721959.1
XM_017024624.1 7761 Intron XP_016880113.1
Gene
RAD51L3-RFFL
Gene Name
RAD51L3-RFFL readthrough
There are no transcripts associated with this gene.

Gene
RFFL
Gene Name
ring finger and FYVE-like domain containing E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017368.1 7761 UTR 3 NP_001017368.1

View Full Product Details