Product Details

SNP ID
rs1135655
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:57979189 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTTAACAGCTTCTTCCCACAGTCT[C/G]AGTGTTTCTACTTGCTTCCCTGGCC
Phenotype
MIM: 606747
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929185 PubMed Links

Gene Details

Gene
LOC101929185
Gene Name
uncharacterized LOC101929185
There are no transcripts associated with this gene.

Gene
VEZF1
Gene Name
vascular endothelial zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007146.2 1448 Silent Mutation CTC,CTG L367L NP_009077.2
XM_005257643.1 1448 Silent Mutation CTC,CTG L373L XP_005257700.1
XM_005257644.4 1448 Silent Mutation CTC,CTG L364L XP_005257701.1
XM_017025017.1 1448 Silent Mutation CTC,CTG L358L XP_016880506.1
XM_017025018.1 1448 Silent Mutation CTC,CTG L358L XP_016880507.1

View Full Product Details