Product Details

SNP ID
rs1052362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21016803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCTACAGTTTATTGTTATAGCA[A/G]AAGTTGTGGGAGACGGGAGGGCACC
Phenotype
MIM: 605272 MIM: 616956
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6717 PubMed Links

Gene Details

Gene
MIR6717
Gene Name
microRNA 6717
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 2171 UTR 3 NP_001269140.1
NM_001282212.1 2171 UTR 3 NP_001269141.1
NM_001282213.1 2171 UTR 3 NP_001269142.1
NM_001282214.1 2171 UTR 3 NP_001269143.1
NM_001282215.1 2171 UTR 3 NP_001269144.1
NM_001282216.1 2171 UTR 3 NP_001269145.1
NM_001320329.1 2171 UTR 3 NP_001307258.1
NM_016250.2 2171 UTR 3 NP_057334.1
NM_201535.1 2171 UTR 3 NP_963293.1
NM_201536.1 2171 UTR 3 NP_963294.1
NM_201537.1 2171 UTR 3 NP_963831.1
NM_201538.1 2171 UTR 3 NP_963832.1
NM_201539.1 2171 UTR 3 NP_963833.1
NM_201540.1 2171 UTR 3 NP_963834.1
NM_201541.1 2171 UTR 3 NP_963835.1
XM_011536996.2 2171 UTR 3 XP_011535298.1
XM_011536997.1 2171 UTR 3 XP_011535299.1
XM_011536998.1 2171 UTR 3 XP_011535300.1
XM_011536999.1 2171 UTR 3 XP_011535301.1
XM_011537001.1 2171 UTR 3 XP_011535303.1
XM_011537002.1 2171 UTR 3 XP_011535304.1
XM_017021480.1 2171 UTR 3 XP_016876969.1
XM_017021481.1 2171 UTR 3 XP_016876970.1
XM_017021482.1 2171 UTR 3 XP_016876971.1
XM_017021483.1 2171 UTR 3 XP_016876972.1
XM_017021484.1 2171 UTR 3 XP_016876973.1
XM_017021485.1 2171 UTR 3 XP_016876974.1
XM_017021486.1 2171 UTR 3 XP_016876975.1
XM_017021487.1 2171 UTR 3 XP_016876976.1
XM_017021488.1 2171 UTR 3 XP_016876977.1
XM_017021489.1 2171 UTR 3 XP_016876978.1
XM_017021490.1 2171 UTR 3 XP_016876979.1
XM_017021491.1 2171 UTR 3 XP_016876980.1
XM_017021492.1 2171 UTR 3 XP_016876981.1
XM_017021493.1 2171 UTR 3 XP_016876982.1
XM_017021494.1 2171 UTR 3 XP_016876983.1
XM_017021495.1 2171 UTR 3 XP_016876984.1
XM_017021496.1 2171 UTR 3 XP_016876985.1
XM_017021497.1 2171 UTR 3 XP_016876986.1
Gene
TPPP2
Gene Name
tubulin polymerization promoting protein family member 2
There are no transcripts associated with this gene.

View Full Product Details