Product Details

SNP ID
rs1951737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23953586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAGAGCTCACCCGCCAGCCCGG[A/G]AAGGCAAGCCCCAGTCAGGCGGAAG
Phenotype
MIM: 611596 MIM: 616925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS4 PubMed Links

Gene Details

Gene
DHRS4
Gene Name
dehydrogenase/reductase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282987.1 Intron NP_001269916.1
NM_001282988.1 Intron NP_001269917.1
NM_001282989.1 Intron NP_001269918.1
NM_001282990.1 Intron NP_001269919.1
NM_001282991.1 Intron NP_001269920.1
NM_021004.3 Intron NP_066284.2
XM_006720005.3 Intron XP_006720068.1
XM_011536366.2 Intron XP_011534668.1
Gene
DHRS4-AS1
Gene Name
DHRS4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details