Product Details
- SNP ID
-
rs967466
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:100547992 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATCAACATTATTACATAAATTAGAC[A/T]GCAGTAAAAATTTCCTAGGGCATAT
- Phenotype
-
MIM: 615276
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CERS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs574236631] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CERS3
- Gene Name
- ceramide synthase 3
There are no transcripts associated with this gene.
- Gene
- PRKXP1
- Gene Name
- protein kinase, X-linked, pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details