Product Details

SNP ID
rs1882527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79028104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCCTGTAGGAATTTTCCTTGAC[A/G]CGAGACAGAGTAGAGCAGGAGACGG
Phenotype
MIM: 610365
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF1 PubMed Links

Gene Details

Gene
C1QTNF1
Gene Name
C1q and tumor necrosis factor related protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030968.3 Intron NP_112230.1
NM_153372.1 Intron NP_699203.1
NM_198593.3 Intron NP_940995.1
NM_198594.2 Intron NP_940996.1
XM_006721663.1 Intron XP_006721726.1
XM_006721664.1 Intron XP_006721727.1
XM_006721666.1 Intron XP_006721729.1
XM_017024126.1 Intron XP_016879615.1
Gene
C1QTNF1-AS1
Gene Name
C1QTNF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details