Product Details

SNP ID
rs1966357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15741413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCCATGCCTGGTCAGAACTACAG[A/C]ACCATATCTGAATTTATCCTCTCTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR10H3 PubMed Links

Gene Details

Gene
OR10H3
Gene Name
olfactory receptor family 10 subfamily H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013938.1 21 Silent Mutation AGA,AGC R7S NP_039226.1

View Full Product Details