Product Details

SNP ID
rs5490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10271151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTGCTACTCAGAGTTGCAACCTC[A/C]GCCTCGCTATGGCTCCCAGCAGCCC
Phenotype
MIM: 147840
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ICAM1 PubMed Links

Gene Details

Gene
ICAM1
Gene Name
intercellular adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000201.2 311 UTR 5 NP_000192.2

View Full Product Details