Product Details

SNP ID
rs1804958
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:157831808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAATGGGGGCTGCTTGTCCCTGA[A/G]AGTAAGGCATAAGCCCAGTGTTCAG
Phenotype
MIM: 602592
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD5L PubMed Links

Gene Details

Gene
CD5L
Gene Name
CD5 molecule like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005894.2 1343 UTR 3 NP_005885.1
XM_005245602.1 1343 Intron XP_005245659.1
XM_017002806.1 1343 Intron XP_016858295.1

View Full Product Details