Product Details

SNP ID
rs172070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18118220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCTCCCGGCCATCGGAGCCGC[C/T]TGGCAAAAGGCGCGCTGCTGCAGCG
Phenotype
MIM: 612258
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAST3 PubMed Links

Gene Details

Gene
MAST3
Gene Name
microtubule associated serine/threonine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015016.1 55 Intron NP_055831.1
XM_005259823.3 55 Silent Mutation CTG,TTG L19L XP_005259880.1
XM_005259824.2 55 Intron XP_005259881.1
XM_005259825.2 55 Intron XP_005259882.1
XM_005259826.3 55 Silent Mutation CTG,TTG L19L XP_005259883.1
XM_005259827.2 55 Silent Mutation CTG,TTG L19L XP_005259884.1
XM_006722698.2 55 Intron XP_006722761.1
XM_006722699.2 55 Intron XP_006722762.2
XM_006722700.3 55 Silent Mutation CTG,TTG L19L XP_006722763.1
XM_011527823.1 55 Intron XP_011526125.1
XM_017026506.1 55 Intron XP_016881995.1
XM_017026507.1 55 Intron XP_016881996.1
XM_017026508.1 55 Intron XP_016881997.1
XM_017026509.1 55 Intron XP_016881998.1
XM_017026510.1 55 Intron XP_016881999.1
XM_017026511.1 55 Intron XP_016882000.1
XM_017026512.1 55 Intron XP_016882001.1
XM_017026513.1 55 Intron XP_016882002.1
XM_017026514.1 55 Intron XP_016882003.1
XM_017026515.1 55 Intron XP_016882004.1
XM_017026516.1 55 Intron XP_016882005.1

View Full Product Details