Product Details
- SNP ID
-
rs2037468
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:27113668 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCATCTTATTATCTGTTTATCAT[A/C]ATAAGAAAATTTGCAATGTCTTTTA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NEK10
PubMed Links
Gene Details
- Gene
- NEK10
- Gene Name
- NIMA related kinase 10
View Full Product Details