Product Details

SNP ID
rs1811974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38039203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGGAAGCCGAAGTGCCGCAGG[G/T]AGTTAGCGGCGTCTCGGTTGCCATG
Phenotype
MIM: 604050 MIM: 602142
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DLEC1 PubMed Links

Gene Details

Gene
DLEC1
Gene Name
deleted in lung and esophageal cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321153.1 608 Intron NP_001308082.1
NM_007335.3 608 Intron NP_031361.2
NM_007337.3 608 Intron NP_031363.2
XM_006713438.3 608 UTR 5 XP_006713501.1
XM_006713439.3 608 UTR 5 XP_006713502.1
XM_006713440.3 608 UTR 5 XP_006713503.1
XM_011534317.2 608 UTR 5 XP_011532619.1
XM_011534318.2 608 UTR 5 XP_011532620.1
Gene
PLCD1
Gene Name
phospholipase C delta 1
There are no transcripts associated with this gene.

View Full Product Details