Product Details

SNP ID
rs1202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86015255 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTCAGAATGGCAGCACCACTGG[C/T]ATGGCGATGGTGCAGGTGGGTGCAG
Phenotype
MIM: 602897
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPK10 PubMed Links

Gene Details

Gene
MAPK10
Gene Name
mitogen-activated protein kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318067.1 3916 UTR 3 NP_001304996.1
NM_001318068.1 3916 UTR 3 NP_001304997.1
NM_001318069.1 3916 UTR 3 NP_001304998.1
NM_002753.4 3916 UTR 3 NP_002744.1
NM_138980.3 3916 UTR 3 NP_620446.1
NM_138982.3 3916 UTR 3 NP_620448.1
XM_005263129.2 3916 UTR 3 XP_005263186.1
XM_005263130.2 3916 UTR 3 XP_005263187.1
XM_005263131.2 3916 UTR 3 XP_005263188.1
XM_005263135.3 3916 UTR 3 XP_005263192.1
XM_006714268.2 3916 UTR 3 XP_006714331.1
XM_006714269.2 3916 UTR 3 XP_006714332.1
XM_011532117.2 3916 UTR 3 XP_011530419.1
XM_011532118.2 3916 UTR 3 XP_011530420.1
XM_011532120.2 3916 UTR 3 XP_011530422.1
XM_011532121.2 3916 UTR 3 XP_011530423.1
XM_017008419.1 3916 UTR 3 XP_016863908.1
XM_017008420.1 3916 UTR 3 XP_016863909.1
XM_017008421.1 3916 UTR 3 XP_016863910.1
XM_017008422.1 3916 UTR 3 XP_016863911.1
XM_017008423.1 3916 UTR 3 XP_016863912.1
XM_017008424.1 3916 UTR 3 XP_016863913.1
XM_017008425.1 3916 UTR 3 XP_016863914.1
XM_017008426.1 3916 UTR 3 XP_016863915.1
XM_017008427.1 3916 UTR 3 XP_016863916.1
XM_017008428.1 3916 UTR 3 XP_016863917.1
XM_017008429.1 3916 UTR 3 XP_016863918.1
XM_017008430.1 3916 UTR 3 XP_016863919.1
XM_017008431.1 3916 UTR 3 XP_016863920.1
XM_017008432.1 3916 UTR 3 XP_016863921.1
XM_017008433.1 3916 UTR 3 XP_016863922.1
XM_017008434.1 3916 UTR 3 XP_016863923.1
XM_017008435.1 3916 UTR 3 XP_016863924.1
XM_017008436.1 3916 UTR 3 XP_016863925.1
XM_017008437.1 3916 UTR 3 XP_016863926.1
XM_017008438.1 3916 UTR 3 XP_016863927.1
XM_017008439.1 3916 UTR 3 XP_016863928.1
XM_017008440.1 3916 UTR 3 XP_016863929.1
XM_017008441.1 3916 UTR 3 XP_016863930.1
XM_017008442.1 3916 UTR 3 XP_016863931.1
XM_017008443.1 3916 UTR 3 XP_016863932.1
XM_017008444.1 3916 UTR 3 XP_016863933.1
XM_017008445.1 3916 UTR 3 XP_016863934.1
XM_017008446.1 3916 UTR 3 XP_016863935.1
XM_017008447.1 3916 UTR 3 XP_016863936.1
XM_017008448.1 3916 UTR 3 XP_016863937.1
XM_017008449.1 3916 UTR 3 XP_016863938.1
XM_017008450.1 3916 UTR 3 XP_016863939.1
XM_017008451.1 3916 UTR 3 XP_016863940.1
XM_017008452.1 3916 UTR 3 XP_016863941.1
XM_017008453.1 3916 Intron XP_016863942.1
XM_017008454.1 3916 Intron XP_016863943.1

View Full Product Details