Product Details
- SNP ID
-
hCV11996198
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:87473765 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGCTGGAAAGTTGAGTTCTTTA[A/T]AATCTTCGTTCAAAACAAGAGATTT
- Phenotype
-
MIM: 606041
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SPARCL1
PubMed Links
Gene Details
- Gene
- SPARCL1
- Gene Name
- SPARC like 1
There are no transcripts associated with this gene.
View Full Product Details