Product Details

SNP ID
rs1997124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:48198745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACTGGGTTTTTTCTCCTGTGCT[A/G]CTAGTTGACTAGAATCTTGTCGCCA
Phenotype
MIM: 607807
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCA13 PubMed Links

Gene Details

Gene
ABCA13
Gene Name
ATP binding cassette subfamily A member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152701.4 Intron NP_689914.3
XM_011515130.2 Intron XP_011513432.1
XM_011515131.2 Intron XP_011513433.1
XM_011515132.2 Intron XP_011513434.1
XM_011515133.2 Intron XP_011513435.1
XM_011515134.2 Intron XP_011513436.1
XM_011515136.2 Intron XP_011513438.2
XM_011515137.2 Intron XP_011513439.1
XM_011515138.2 Intron XP_011513440.1
XM_011515139.2 Intron XP_011513441.1
XM_011515141.2 Intron XP_011513443.1
XM_011515142.2 Intron XP_011513444.1
XM_011515143.2 Intron XP_011513445.1
XM_011515144.2 Intron XP_011513446.1
XM_011515145.2 Intron XP_011513447.1
XM_011515146.2 Intron XP_011513448.1
XM_011515147.2 Intron XP_011513449.1
XM_011515148.2 Intron XP_011513450.1
XM_011515149.2 Intron XP_011513451.1
XM_017011767.1 Intron XP_016867256.1
XM_017011768.1 Intron XP_016867257.1
XM_017011769.1 Intron XP_016867258.1

View Full Product Details