Product Details

SNP ID
rs6563
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:136494732 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGGCTAAGGCTCCCCGAGCTGAGCC[A/G]AGTCTGACGTCCCTCACTGGCATGA
Phenotype
MIM: 190198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf163 PubMed Links

Gene Details

Gene
C9orf163
Gene Name
chromosome 9 open reading frame 163
There are no transcripts associated with this gene.

Gene
NOTCH1
Gene Name
notch 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017617.4 9007 UTR 3 NP_060087.3
XM_011518717.2 9007 Intron XP_011517019.2

View Full Product Details