Product Details

SNP ID
rs475215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57556715 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCCTATCAGTCAGAGAAGGCC[A/C]ACAGCTGAGGCCTCAGAGCAGGATG
Phenotype
MIM: 613664 MIM: 603890
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
There are no transcripts associated with this gene.

Gene
UBE2L6
Gene Name
ubiquitin conjugating enzyme E2 L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004223.4 Intron NP_004214.1
NM_198183.2 Intron NP_937826.1

View Full Product Details