Product Details
- SNP ID
-
rs14341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:49431698 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGATCATAACTAAAATAATATT[C/T]TAGACAAAAGCTAAAACAAAAAGAG
- Phenotype
-
MIM: 609058
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MUT
PubMed Links
Gene Details
- Gene
- MUT
- Gene Name
- methylmalonyl-CoA mutase
View Full Product Details