Product Details

SNP ID
rs121918252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49431831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGACCAAATACATTGGAAACA[A/C]CAACTTCAAACAGAAATTCATAATC
Phenotype
MIM: 609058
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MUT PubMed Links

Gene Details

Gene
MUT
Gene Name
methylmalonyl-CoA mutase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000255.3 2415 Missense Mutation GGT,GTT G717V NP_000246.2
XM_005249143.3 2415 Missense Mutation GGT,GTT G717V XP_005249200.1

View Full Product Details