Product Details

SNP ID
rs12798364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:70072609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAGGATTCGCCCCAGAGTCAGA[A/G]GCTTGCTGCAGCCAGTGCCTGTAAC
Phenotype
MIM: 610108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANO1 PubMed Links

Gene Details

Gene
ANO1
Gene Name
anoctamin 1
There are no transcripts associated with this gene.

Gene
ANO1-AS2
Gene Name
ANO1 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

Gene
LOC101928443
Gene Name
uncharacterized LOC101928443
There are no transcripts associated with this gene.

View Full Product Details