Product Details

SNP ID
rs14339
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:9848530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGGCAGCCAGCAATGTGGCCT[C/T]GACACCTGACTCCAGGCACTCCGAC
Phenotype
MIM: 607758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNBIP1 PubMed Links

Gene Details

Gene
CTNNBIP1
Gene Name
catenin beta interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012329.1 2966 UTR 3 NP_001012329.1
NM_020248.2 2966 UTR 3 NP_064633.1
XM_017001853.1 2966 UTR 3 XP_016857342.1
XM_017001854.1 2966 UTR 3 XP_016857343.1
XM_017001855.1 2966 UTR 3 XP_016857344.1

View Full Product Details