Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033046.3 | 1935 | UTR 3 | NP_001028218.1 | ||
NM_001100407.2 | 1935 | UTR 3 | NP_001093877.1 | ||
NM_001100408.2 | 1935 | UTR 3 | NP_001093878.1 | ||
NM_001193653.1 | 1935 | UTR 3 | NP_001180582.1 | ||
NM_001193654.1 | 1935 | UTR 3 | NP_001180583.1 | ||
NM_001193655.1 | 1935 | UTR 3 | NP_001180584.1 | ||
NM_001193657.1 | 1935 | UTR 3 | NP_001180586.1 | ||
XM_006722293.2 | 1935 | UTR 3 | XP_006722356.1 | ||
XM_011523606.2 | 1935 | UTR 3 | XP_011521908.1 | ||
XM_017025072.1 | 1935 | UTR 3 | XP_016880561.1 | ||
XM_017025073.1 | 1935 | UTR 3 | XP_016880562.1 | ||
XM_017025074.1 | 1935 | UTR 3 | XP_016880563.1 | ||
XM_017025075.1 | 1935 | UTR 3 | XP_016880564.1 | ||
XM_017025076.1 | 1935 | UTR 3 | XP_016880565.1 | ||
XM_017025077.1 | 1935 | UTR 3 | XP_016880566.1 | ||
XM_017025078.1 | 1935 | UTR 3 | XP_016880567.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173620.2 | 1935 | Intron | NP_775891.2 | ||
XM_005256346.3 | 1935 | Intron | XP_005256403.1 | ||
XM_005256347.4 | 1935 | Intron | XP_005256404.1 | ||
XM_006722280.3 | 1935 | Intron | XP_006722343.1 | ||
XM_011523560.2 | 1935 | Intron | XP_011521862.1 | ||
XM_011523561.2 | 1935 | Intron | XP_011521863.1 | ||
XM_017024484.1 | 1935 | Intron | XP_016879973.1 | ||
XM_017024485.1 | 1935 | Intron | XP_016879974.1 |