Product Details

SNP ID
rs2043624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:29503671 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAACTCTGATCATTGGTTCAA[A/C]TGTTCAGCCAGGCCAAAGCAACTCC
Phenotype
MIM: 615855
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OVCH1 PubMed Links

Gene Details

Gene
OVCH1
Gene Name
ovochymase 1
There are no transcripts associated with this gene.

Gene
TMTC1
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193451.1 6135 UTR 3 NP_001180380.1
NM_175861.3 6135 UTR 3 NP_787057.2
XM_005253499.4 6135 UTR 3 XP_005253556.1
XM_017020003.1 6135 UTR 3 XP_016875492.1
XM_017020004.1 6135 UTR 3 XP_016875493.1
XM_017020005.1 6135 UTR 3 XP_016875494.1
XM_017020006.1 6135 Intron XP_016875495.1
XM_017020007.1 6135 UTR 3 XP_016875496.1

View Full Product Details