Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020371.2 | Intron | NP_065104.1 | |||
XM_011521818.2 | Intron | XP_011520120.1 | |||
XM_011521819.2 | Intron | XP_011520121.1 | |||
XM_011521820.1 | Intron | XP_011520122.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001036.4 | Intron | NP_001027.3 | |||
NM_001243996.2 | Intron | NP_001230925.1 | |||
XM_011521880.2 | Intron | XP_011520182.1 | |||
XM_017022468.1 | Intron | XP_016877957.1 | |||
XM_017022469.1 | Intron | XP_016877958.1 | |||
XM_017022470.1 | Intron | XP_016877959.1 | |||
XM_017022471.1 | Intron | XP_016877960.1 | |||
XM_017022472.1 | Intron | XP_016877961.1 | |||
XM_017022473.1 | Intron | XP_016877962.1 | |||
XM_017022474.1 | Intron | XP_016877963.1 | |||
XM_017022475.1 | Intron | XP_016877964.1 | |||
XM_017022476.1 | Intron | XP_016877965.1 | |||
XM_017022477.1 | Intron | XP_016877966.1 |