Product Details

SNP ID
rs1878188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48138226 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAACATTAGTTTTAACATGATTT[C/G]ATCCAATTGTTACATTATTTTGTAA
Phenotype
MIM: 609802
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYEF2 PubMed Links

Gene Details

Gene
MYEF2
Gene Name
myelin expression factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301210.1 6433 Intron NP_001288139.1
NM_016132.4 6433 Intron NP_057216.2
XM_005254422.4 6433 Intron XP_005254479.2
XM_005254424.4 6433 Intron XP_005254481.2
XM_005254425.4 6433 UTR 3 XP_005254482.2
XM_005254427.4 6433 Intron XP_005254484.1
XM_006720553.3 6433 Intron XP_006720616.1
XM_011521657.2 6433 Intron XP_011519959.1
XM_017022285.1 6433 UTR 3 XP_016877774.1
XM_017022286.1 6433 UTR 3 XP_016877775.1
XM_017022287.1 6433 UTR 3 XP_016877776.1
XM_017022288.1 6433 Intron XP_016877777.1
XM_017022289.1 6433 Intron XP_016877778.1
XM_017022290.1 6433 Intron XP_016877779.1
XM_017022291.1 6433 UTR 3 XP_016877780.1
XM_017022292.1 6433 UTR 3 XP_016877781.1
Gene
SLC24A5
Gene Name
solute carrier family 24 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205850.2 6433 Intron NP_995322.1
XM_017022079.1 6433 Intron XP_016877568.1
XM_017022080.1 6433 Intron XP_016877569.1

View Full Product Details