Product Details

SNP ID
rs10790371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:120240332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGGGGCGCTGGCTTTGGCCCC[C/G]CCTGGGGCAGGATGGTGAATCTGGA
Phenotype
MIM: 607394
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC649133 PubMed Links

Gene Details

Gene
LOC649133
Gene Name
uncharacterized LOC649133
There are no transcripts associated with this gene.

Gene
POU2F3
Gene Name
POU class 2 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244682.1 139 Intron NP_001231611.1
NM_014352.3 139 UTR 5 NP_055167.2
XM_011542739.2 139 UTR 5 XP_011541041.1
XM_011542740.2 139 UTR 5 XP_011541042.1
XM_011542741.2 139 UTR 5 XP_011541043.1
XM_011542742.2 139 UTR 5 XP_011541044.1
XM_011542743.2 139 Intron XP_011541045.1
XM_017017487.1 139 UTR 5 XP_016872976.1

View Full Product Details